missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Electrolyte For Cyanide ISE
€ 35.00 - € 46.70
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11977721
|
Mettler Toledo™
51107893 |
20 mL |
€ 46.70
20mL |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
12714444
|
Mettler Toledo™
51107893 |
20 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Electrolyte | |
20 mL |