Learn More
SI Analytics™ Electrolyte Solution for Ammonia Electrodes
For use with ammonia combination electrodes
Brand: SI Analytics™ 285137344
Additional Details : Weight : 0.25000kg
Description
- Plastic bottle
- Type L 6408
Specifications
For Combination Ammonia Electrodes | |
Electrolyte solution | |
50 mL |
L 6408 | |
Plastic bottle |