missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF1 Monoclonal Antibody (OTI4F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811197S
This item is not returnable.
View return policy
Description
ELF1 Monoclonal antibody specifically detects ELF1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ELF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UH03, Q9Z1S5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 24050, 55964 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4F4 | |
| Unconjugated | |
| SEPT3 | |
| E74 like ETS transcription factor 1; E74-like factor 1; E74-like factor 1 (ets domain transcription factor); ELF 1; ELF1; Elf-1; ETS-like transcription factor Elf-1-like protein; ETS-related transcription factor Elf-1; ETS-related transcription factor elf-1-like protein; mElf 1; mElf-1; OTTHUMP00000018310; p70; Sts1 | |
| Full length human recombinant protein of human SEPT3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction