missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585556
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (83%), Rat (80%), Rhesus Monkey (99%), Bovine (81%). Positive Control: HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants.
Specifications
| ELF1 | |
| Polyclonal | |
| Unconjugated | |
| ELF1 | |
| E74 like ETS transcription factor 1; E74-like factor 1; E74-like factor 1 (ets domain transcription factor); ELF 1; ELF1; Elf-1; ETS-like transcription factor Elf-1-like protein; ETS-related transcription factor Elf-1; ETS-related transcription factor elf-1-like protein; mElf 1; mElf-1; OTTHUMP00000018310; p70; Sts1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1997 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P32519 | |
| ELF1 | |
| Recombinant protein encompassing a sequence within the center region of human ELF1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction