missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF3 Monoclonal Antibody (OTI7H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810005
This item is not returnable.
View return policy
Description
ESE-1 (Epithelium-Specific Ets) is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family, which has features distinct from those of any other ets-related factor. ESE-1 binds with high affinity to and transactivates the ets binding site in the promoter of the keratinocyte terminal differentiation marker gene, SPRR2A. ESE-1 is a critical regulator of epithelial cell differentiation.Specifications
| ELF3 | |
| Monoclonal | |
| Unconjugated | |
| ELF3 | |
| EPR-1, ERT, ESE-1, ESX | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1999 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI7H9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78545 | |
| ELF3 | |
| Human recombit protein fragment corresponding to amino acids 175-286 of human ELF3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction