missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF3 Monoclonal Antibody (OTI7H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810005S
This item is not returnable.
View return policy
Description
ELF3 Monoclonal antibody specifically detects ELF3 in Human samples. It is validated for Western BlotSpecifications
| ELF3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P83369 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 134353 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI7H9 | |
| Unconjugated | |
| LSM11 | |
| E74 like ETS transcription factor 3; E74-like factor 3; E74-like factor 3 (ets domain transcription factor); E74-like factor 3 (ets domain transcription factor, epithelial-specific ); E74-like factor 3 (ETS domain transcription factor, serine box, epithelial-specific); ELF3; elf3 {ECO:0000312; EMBL:AAH97980.1}; Epithelial-restricted with serine box; epithelium-restricted Ets protein ESX; Epithelium-specific Ets transcription factor 1; EPR-1; Ert; ESE-1; ESX; ets domain transcription factor, serine box (epithelial-specific); ETS-related transcription factor Elf-3; JEN; RP11-510N19.1 | |
| Full length human recombinant protein of human LSM11 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction