missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELF3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16517221
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16517221 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16517221 Supplier Invitrogen™ Supplier No. PA589261

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAATCEISNI FSNYFSAMYS SEDSTLASVP PAATFGADDL VLTLSNPQMS LEGTEKASWL GEQPQFWSKT QVLDWISYQV EKNKYDASAI DFSRCDMDGA TLCNCALEEL RLVFGPLGDQ LHAQLRDLTS SSSDELSWII ELLEKDGMAF QEALDPGPFD QGSPFAQELL DDGQQASPYH PGSCGAGAPS PGSSDVSTAG TGASRSSHSS DSGGSDVDLD PTDGKLFPSD GFRDCKKGDP KHGKRKRGRP RKLSKEYWDC LEGKKSKHAP RGTHLWEFIR; Positive Samples: Mouse skin, Mouse lung, SGC-7901, HT-29, SW480; Cellular Location: Cytoplasm, Nucleus.

ESE-1 (Epithelium-Specific Ets) is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family, which has features distinct from those of any other ets-related factor. ESE-1 binds with high affinity to and transactivates the ets binding site in the promoter of the keratinocyte terminal differentiation marker gene, SPRR2A. ESE-1 is a critical regulator of epithelial cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELF3
Applications ELISA, Immunoprecipitation, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ELF3
Gene Accession No. P78545, Q3UPW2
Gene Alias E74 like ETS transcription factor 3; E74-like factor 3; E74-like factor 3 (ets domain transcription factor); E74-like factor 3 (ets domain transcription factor, epithelial-specific ); E74-like factor 3 (ETS domain transcription factor, serine box, epithelial-specific); ELF3; elf3 {ECO:0000312; EMBL:AAH97980.1}; Epithelial-restricted with serine box; epithelium-restricted Ets protein ESX; Epithelium-specific Ets transcription factor 1; EPR-1; Ert; ESE-1; ESX; ets domain transcription factor, serine box (epithelial-specific); ETS-related transcription factor Elf-3; JEN; RP11-510N19.1
Gene Symbols ELF3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ESE1/ESE1/ELF3 (NP_0044243).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13710, 1999
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.