missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elf4/MEF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16314
This item is not returnable.
View return policy
Description
Elf4/MEF Polyclonal specifically detects Elf4/MEF in Human samples. It is validated for Western Blot.
Specifications
| Elf4/MEF | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| E74-like factor 4, E74-like factor 4 (ets domain transcription factor), ETS-related transcription factor Elf-4, MEFELFRMyeloid Elf-1-like factor | |
| Rabbit | |
| 71 kDa | |
| 0.1 mL | |
| Cancer | |
| 2000 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q99607 | |
| ELF4 | |
| Recombinant protein encompassing a sequence within the center region of human ELF4. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (80%), Rat (81%), Bovine (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction