missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF4 Monoclonal Antibody (OTI4D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810142
This item is not returnable.
View return policy
Description
ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5′-WGGA-3′. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.Specifications
| ELF4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99607 | |
| ELF4 | |
| Human recombit protein fragment corresponding to amino acids 83-345 of human ELF4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4D5 | |
| Unconjugated | |
| ELF4 | |
| ELFR, MEF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2000 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction