Learn More
ELF4 Mouse anti-Human, Clone: OTI2D8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810090
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5′-WGGA-3′. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.Specifications
| ELF4 | |
| Monoclonal | |
| Unconjugated | |
| ELF4 | |
| ELFR, MEF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2000 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q99607 | |
| ELF4 | |
| Human recombit protein fragment corresponding to amino acids 83-345 of human ELF4 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.