missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELFN1 Antibody (965705) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10644JF585
This item is not returnable.
View return policy
Description
ELFN1 Monoclonal antibody specifically detects ELFN1 in Human samples. It is validated for Flow Cytometry
Specifications
| ELFN1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| USP17L7 | |
| Chinese Hamster Ovary cell line, CHO-derived human ELFN1 protein, Asp28-Tyr418, Accession # P0C7U0 | |
| RUO | |
| 392617 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 965705 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction