Learn More
Invitrogen™ Elipovimab Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542239
Product Code. 17718863
  • € 524.00 / 100µg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Elipovimab (formerly GS-9722;PGT121-derived broadly neutralizing antibody).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Elipovimab
Recombinant Monoclonal
Unconjugated
GS-HIV
Human HIV-1 gp120.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, SDS-Page
2.05 mg/mL
PBS with no preservative; pH 7.4
Human
Protein A
RUO
Virus
Antibody
IgG1 λ