missing translation for 'onlineSavingsMsg'
Learn More

Eliprodil, MedChemExpress

Product Code. p-200101345
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16405438

Brand: Medchem Express HY12881

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eliprodil(SL-820715) is a non-competitive NR2B-NMDA receptor antagonist(IC50=1 uM), less potent for NR2A- and NR2C-containing receptors(IC50> 100 uM).

Eliprodil(SL-820715) is a non-competitive NR2B-NMDA receptor antagonist(IC50=1 uM), less potent for NR2A- and NR2C-containing receptors(IC50> 100 uM).

Specifications

Chemical Name or Material Eliprodil
CAS 119431-25-3
Color White
Purity Grade Notes Research
Molecular Formula C20H23ClFNO
Quantity 10 mg
Synonym SL-820715
Solubility Information DMSO : 14.29 mg/mL (41.08 mM; Need ultrasonic)
SMILES ClC(C=C1)=CC=C1C(O)CN(CC2)CCC2CC3=CC=C(F)C=C3
Molecular Weight (g/mol) 347.85
Formula Weight 347.85
Percent Purity 98.36%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.