missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1 Antibody (PCRP-ELK1-1D9), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08888V
This item is not returnable.
View return policy
Description
Elk-1 Monoclonal antibody specifically detects Elk-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| Elk-1 | |
| Monoclonal | |
| DyLight 405 | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| Recombinant full-length human Elk-1 protein (Uniprot: P19419) | |
| 100 μg | |
| Autophagy, Cancer, MAP Kinase Signaling, Neuroscience | |
| 2002 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunofluorescence, Protein Array | |
| PCRP-ELK1-1D9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction