missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1 Mouse anti-Human, Clone: PCRP-ELK1-1B9, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07142-100ug
This item is not returnable.
View return policy
Description
Elk-1 Monoclonal specifically detects Elk-1 in Human samples. It is validated for ELISA, Immunoprecipitation, Protein Array.
Specifications
| Elk-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein 1 mL of lysate, Protein Array | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| Recombinant full-length human Elk-1 protein | |
| 100 μg | |
| Autophagy, Cancer, MAP Kinase Signaling, Neuroscience, Phospho Specific | |
| 2002 | |
| Store at 4°C. | |
| IgG2a |
| ELISA, Immunoprecipitation, Peptide Array | |
| PCRP-ELK1-1B9 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction