missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1, p Ser383 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 479.00
Specifications
| Antigen | Elk-1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639386
|
Novus Biologicals
NBP3-05380-100ul |
100 μg |
€ 479.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695058
|
Novus Biologicals
NBP3-05380-20ul |
20 μg |
€ 201.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Elk-1 Polyclonal antibody specifically detects Elk-1 in Human samples. It is validated for Western BlotSpecifications
| Elk-1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, MAP Kinase Signaling, Neuroscience, Phospho Specific | |
| PBS with 50% glycerol, pH7.3. | |
| 2002 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| A synthetic phosphorylated peptide around S383 of human ELK1 (NP_005220.2). TLSPI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title