missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1, p Ser389 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54759FR
This item is not returnable.
View return policy
Description
Elk-1 Polyclonal antibody specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Elk-1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| Elk1 (Phospho-Ser389) antibody was raised against a peptide sequence around phosphorylation site of serine 389 (P-R-S (p) -P-A) derived from Human Elk-1. | |
| 0.1 mL | |
| Autophagy, Cancer, MAP Kinase Signaling, Neuroscience, Phospho Specific | |
| 2002 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Epitope affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction