missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1, p Ser389 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54759-0.05MG
This item is not returnable.
View return policy
Description
Elk-1 Polyclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Elk-1 | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, and 50% glycerol. with 0.02% sodium azide | |
| ELK1 | |
| Elk1 (Phospho-Ser389) antibody was raised against a peptide sequence around phosphorylation site of serine 389 (P-R-S (p) -P-A) derived from Human Elk-1. | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:10-1:500 | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| Rabbit | |
| 0.05 mg | |
| 2002 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction