missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587504
This item is not returnable.
View return policy
Description
Immunogen sequence: PSPLEACLEA EEAGLPLQVI LTPPEAPNLK SEELNVEPGL GRALPPEVKV EGPKEELEVA GERGFVPETT KAEPEVPPQE GVPARLPAVV MDTAGQA Positive Samples: HeLa, Jurkat, HT-29, K-562, Mouse liver, Rat brain; Cellular Location: Nucleus.
ELK1 is a component of the ternary complex that binds the serum response element (SRE) and mediates gene activity in response to serum and growth factors. ELK1 is phosphorylated by MAP kinase pathways at a cluster of S/T motifs at its C-terminus. Phosphorylation at these sites, particularly Ser383, is critical for transcriptional activation by ELK1. ELK1 appears to be a direct target of activated MAP kinase. Biochemical studies indicate that ELK1 is a good substrate for MAP kinase, the kinetics of ELK1 phosphorylation and activation correlate with MAP kinase activity, and interfering mutants of MAP kinase block ELK1 activation in vivo. ELK1 is a nuclear target for the ras-raf-MAPK signaling cascade. Alternatively spliced transcript variants encoding the same protein have been found for ELK1.
Specifications
| ELK1 | |
| Polyclonal | |
| Unconjugated | |
| ELK1 | |
| ELK 1; ELK1; Elk-1; ELK1, ETS transcription factor; ELK1, member of ETS oncogene family; ETS domain-containing protein Elk-1; ETS transcription factor ELK1; ETS-like gene 1; Oncogene Elk1; tyrosine kinase (ELK1) oncogene | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13712, 2002, 314436 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A4GTP4, P19419, P41969 | |
| ELK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 201-297 of human ELK1 (NP_0052202). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction