missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELK3 Monoclonal Antibody (OTI2C10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503602
This item is not returnable.
View return policy
Description
May be a negative regulator of transcription, but can activate transcription when coexpressed with Ras, Src or Mos. Forms a ternary complex with the serum response factor and the ETS and SRF motifs of the Fos serum response element.Specifications
| ELK3 | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41970 | |
| ELK3 | |
| Full length human recombit protein of human ELK3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C10 | |
| Unconjugated | |
| ELK3 | |
| ERP, NET, SAP-2, SAP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2004 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction