missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELK3 Monoclonal Antibody (OTI4A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503559S
This item is not returnable.
View return policy
Description
ELK3 Monoclonal antibody specifically detects ELK3 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| ELK3 | |
| Monoclonal | |
| 0.77 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P28070 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5692 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI4A4 | |
| Unconjugated | |
| PSMB4 | |
| D430049E23Rik; ELK3; ELK3, ETS transcription factor; ELK3, ETS-domain protein; ELK3, ETS-domain protein (SRF accessory protein 2); ELK3, member of ETS oncogene family; Erp; Etrp; ETS domain-containing protein Elk-3; ETS transcription factor ELK3; ETS-domain protein ELK-3; ETS-related protein ERP; ETS-related protein NET; Net; SAP2; Sap-2; Serum response factor accessory protein 2; si:ch211-172b12.2; si:dkey-201e17.1; SRF accessory protein 2; zgc:198153; zgc:198183 | |
| Full length human recombinant protein of human PSMB4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction