missing translation for 'onlineSavingsMsg'
Learn More

ELK3 Monoclonal Antibody (OTI4G9), TrueMAB™, OriGene

Product Code. p-200011336
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17589571 30 μL 30µL
17569571 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17589571 Supplier Origene Technologies Supplier No. TA503575S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ELK3 Monoclonal antibody specifically detects ELK3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ELK3
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FABP2
Gene Accession No. P12104
Gene Alias D430049E23Rik; ELK3; ELK3, ETS transcription factor; ELK3, ETS-domain protein; ELK3, ETS-domain protein (SRF accessory protein 2); ELK3, member of ETS oncogene family; Erp; Etrp; ETS domain-containing protein Elk-3; ETS transcription factor ELK3; ETS-domain protein ELK-3; ETS-related protein ERP; ETS-related protein NET; Net; SAP2; Sap-2; Serum response factor accessory protein 2; si:ch211-172b12.2; si:dkey-201e17.1; SRF accessory protein 2; zgc:198153; zgc:198183
Host Species Mouse
Immunogen Full length human recombinant protein of human FABP2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2169
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.