missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELL Monoclonal Antibody (OTI2H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810799S
This item is not returnable.
View return policy
Description
ELL Monoclonal antibody specifically detects ELL in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ELL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P55199 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8178 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2H7 | |
| Unconjugated | |
| Ell | |
| C19orf17; Eleven-nineteen lysine-rich leukemia protein; Ell; ELL gene (11-19 lysine-rich leukemia gene); ELL1; elongation factor for RNA polymerase II; elongation factor RNA polymerase II; hypothetical protein LOC325221; MEN; PPP1R68; protein phosphatase 1, regulatory subunit 68; RNA polymerase II elongation factor ELL; RNA POLYMERASE II ELONGATION FACTOR ELL (ELEVEN-NINETEEN LYSINE-RICH LEUKEMIA PROTEIN); wu:fc60b11; zgc:63836 | |
| Human recombinant protein fragment corresponding to amino acids 302-621 of human ELL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction