missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELL Monoclonal Antibody (OTI2H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810799
This item is not returnable.
View return policy
Description
Elongation factor that can increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA.Specifications
| ELL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P55199 | |
| ELL | |
| Human recombit protein fragment corresponding to amino acids 302-621 of human ELL produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2H7 | |
| Unconjugated | |
| ELL | |
| C19orf17, ELL1, MEN, PPP1R68 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8178 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction