Learn More
ELL Monoclonal Antibody (OTI4C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810844
This item is not returnable.
View return policy
Description
Elongation factor that can increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA.Specifications
| ELL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P55199 | |
| ELL | |
| Human recombit protein fragment corresponding to amino acids 302-621 of human ELL produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4C4 | |
| Unconjugated | |
| ELL | |
| C19orf17, ELL1, MEN, PPP1R68 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8178 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |