Learn More
Invitrogen™ ELL2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5100683
Product Code. 16667164
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total ELL2.
SHP-1 (PTPN6) is a non-receptor protein tyrosine phosphatase that is expressed primarily in hematopoietic cells. The enzyme is composed of two SH2 domains, a tyrosine phosphatase catalytic domain, and a carboxy-terminal regulatory domain. SHP-1 removes phosphates from target proteins to downregulate several tyrosine kinase-regulated pathways. In hematopoietic cells, the amino-terminal SH2 domain of SHP-1 binds to tyrosine phosphorylated erythropoietin receptors (EpoR) to negatively regulate hematopoietic growth. Overexpression of SHP-1 in epithelial cells results in dephosphorylation of the Ros receptor tyrosine kinase and subsequent downregulation of Ros-dependent cell proliferation and transformation. Following ligand binding in myeloid cells, SHP-1 associates with the IL-3R beta chain and downregulates IL-3-induced tyrosine phosphorylation and cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELL2
Polyclonal
Unconjugated
ELL2
cb244; eleven-nineteen lysine-rich leukemia gene 2; ELL2; ELL-related RNA polymerase II, elongation factor; elongation factor for RNA polymerase II 2; elongation factor RNA for polymerase II 2; elongation factor RNA polymerase II 2; elongation factor, RNA polymerase II 2; elongation factor, RNA polymerase II, 2; fb78f03; im:3814960; RNA polymerase II elongation factor ELL2; RNA polymerase II elongation factor ELL2-like protein; sb:cb244; wu:fb78f03; wu:fb78f03 protein
Rabbit
Affinity chromatography
RUO
192657, 22936
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O00472, Q3UKU1
ELL2
A synthesized peptide derived from human ELL2(Accession O00472), corresponding to amino acid residues G161-I211.
100 μL
Primary
Human, Mouse