Learn More
Bel-Art™ Ellipso-Spoon and Spatula Samplers
Product Code. 15510132 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
10 mL Bowl
70 mL bowl
Length (Metric):
15 cm
21 cm
30 cm
40 cm
50 cm
Size:
15 x 35 mm bowl
28 x 65 mm bowl
Unit Size:
Each
Product Code. 15510132
Brand: Bel-Art™ H368080030
Only null left
Add to basket
Add to basket
Special curved shape prevents missed sample accumulation in corners.
  • Crystals, powders, granulated and solid substances, and thick liquids retrieved
  • Includes spoon end and spatula end
  • Made of polished 304 grade stainless steel
Specifications
Autoclavable Autoclavable
Capacity (Metric) 70 mL bowl
Product Type Spoon and Spatula Sampler
Finish Polished
For Use With (Equipment) Bottles and containers
For Use With (Application) Designed to extracts crystals, granulated and solid substances, thick liquids and delicate materials
Size 28 x 65 mm bowl
Material Stainless Steel
Length (Metric) 30 cm