missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELMO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589438
This item is not returnable.
View return policy
Description
Immunogen sequence: SLPNPEYYTL RYADGPQLYI TEQTRSDIKN GTILQLAISP SRAARQLMER TQSSNMETRL DAMKELAKLS ADVTFATEF; Positive Samples: HeLa, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Membrane, cytosol.
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein. Similarity to a C. elegans protein suggests that this protein may function in phagocytosis of apoptotic cells and in cell migration. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| ELMO2 | |
| Polyclonal | |
| Unconjugated | |
| ELMO2 | |
| 1190002F24Rik; CED12; CED-12; ced-12 homolog 2; CED-12 homolog A; CED12A; Ced-12A; Elmo2; ELMO-2; engulfment and cell motility 2; engulfment and cell motility 2, ced-12 homolog; engulfment and cell motility protein 2; FLJ11656; hCed-12A; KIAA1834; PH domain protein CED12A; protein ced-12 homolog A; RP11-394O2.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140579, 362271, 63916 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8BHL5, Q96JJ3 | |
| ELMO2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 42-120 of human ELMO2 (NP_8774961). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction