missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELMO3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577159
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene is similar to a C. elegans protein that functions in phagocytosis of apoptotic cells and in cell migration. Other members of this small family of engulfment and cell motility (ELMO) proteins have been shown to interact with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes.
Specifications
| ELMO3 | |
| Polyclonal | |
| Unconjugated | |
| ELMO3 | |
| 9930107J06; BC058752; Ced12; CED-12; ced-12 homolog 3; Elmo3; ELMO-3; engulfment and cell motility 3; engulfment and cell motility 3, ced-12 homolog; engulfment and cell motility protein 3; FLJ13824 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 234683, 291962, 79767 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q499U2, Q8BYZ7, Q96BJ8 | |
| ELMO3 | |
| Recombinant full length Human ELMO3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction