missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELOA Polyclonal Antibody

Product Code. 15693855
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15693855 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15693855 Supplier Invitrogen™ Supplier No. PA549928

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total Elongin A protein.

This gene encodes the transcriptionally active subunit of the SIII (or elongin) transcription elongation factor complex, which also includes two regulatory subunits, elongins B and C. This complex acts to increase the rate of RNA chain elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites along the DNA template. Whereas a related protein with similar function, elongin A, is ubiquitously expressed, the encoded protein is specifically expressed in the testis, suggesting it may have a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ELOA
Gene Accession No. Q14241, Q63187, Q8CB77, Q8IYF1
Gene Alias 110kDa; AA408125; El; EloA; EloA2; Elongin 110 kDa subunit; elongin A; Elongin A2; elongin-A; Elongin-A2; MSTP059; RNA polymerase II transcription factor elongin subunit A (110 kDa); RNA polymerase II transcription factor SIII subunit A; RNA polymerase II transcription factor SIII subunit A1; SIII; SIII p110; TCEB3; TCEB3A; TCEB3B; TCEB3L; transcription elongation factor B (SIII), polypeptide 3; transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A); transcription elongation factor B alpha subunit; transcription elongation factor B polypeptide 3; transcription elongation factor B subunit 3
Gene Symbols ELOA, ELOA2
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human ELOA1 (aa 131-180).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25562, 27224, 51224, 6924
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.