missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elongin A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-40834
This item is not returnable.
View return policy
Description
Elongin A Polyclonal specifically detects Elongin A in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Elongin A | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| EloA, Elongin 110 kDa subunit, elongin-A, FLJ38760, FLJ42849, RNA polymerase II transcription factor SIII subunit A1, SIII, SIII p110, TCEB3A, transcription elongation factor B (SIII), polypeptide 3 (110kD, elongin A), transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A), transcription elongation factor B alpha subunit, transcription elongation factor B polypeptide 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6924 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| NP_003189.1 | |
| TCEB3 | |
| The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human Transcription Elongation Factor B (SIII), Polypeptide 3 (110kDa, elongin A) using the numbering given in entry NP_003189.1 (GeneID 6924). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction