missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL3 Antibody (V61 P2B3*D10), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50385DL594
This item is not returnable.
View return policy
Description
ELOVL3 Monoclonal antibody specifically detects ELOVL3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| ELOVL3 | |
| Monoclonal | |
| DyLight 594 | |
| 3-keto acyl-CoA synthase ELOVL3, CIG-30, CIG30MGC21435, Cold-inducible glycoprotein of 30 kDa, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3, elongation of very long chain fatty acids protein 3 | |
| Ovalbumin-conjugated synthetic peptide - QPYNFELSK | |
| 0.1 mL | |
| Cancer, Cellular Signaling, metabolism | |
| 83401 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| V61 P2B3*D10 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction