missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELOVL3 Monoclonal Antibody (V61P2B3-D10), PerCP

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545908
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of MA5-45908 was sufficient for detection of Elongation of very long chain fatty acids protein 3 in 10 μg of Rat Brain by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects 40 kDa.
This gene encodes a protein that belongs to the GNS1/SUR4 family. Members of this family play a role in elongation of long chain fatty acids to provide precursors for synthesis of sphingolipids and ceramides. [provided by RefSeq, Jul 2013].
Specifications
| ELOVL3 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q9HB03 | |
| ELOVL3 | |
| Synthetic peptide of Human Elongation of very long chain fatty acids protein 3. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| V61P2B3-D10 | |
| PerCP | |
| ELOVL3 | |
| 3-keto acyl-CoA synthase ELOVL3; CIG30; CIG-30; Cold-inducible glycoprotein of 30 kDa; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3; elongation of very long chain fatty acids protein 3; ELOVL FA elongase 3; ELOVL fatty acid elongase 3; ELOVL3; Very long chain 3-ketoacyl-CoA synthase 3; very long chain 3-oxoacyl-CoA synthase 3; very-long-chain 3-oxoacyl-CoA synthase 3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 309449, 83401 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction