missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL7 Antibody (Z26), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50384UV
This item is not returnable.
View return policy
Description
ELOVL7 Monoclonal specifically detects ELOVL7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| ELOVL7 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ELOVL7 | |
| Peptide Sequence - RAYTKGQRLP (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), bovine (Bos taurus), canine (Canis familiaris), equine (Equus caballus) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 79993 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Z26 | |
| Western Blot, ELISA, Immunohistochemistry | |
| ELOVL family member 7, elongation of long chain fatty acids, ELOVL fatty acid elongase 7 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction