missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELP4 Monoclonal Antibody (OTI3A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809150
This item is not returnable.
View return policy
Description
This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.Specifications
| ELP4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96EB1 | |
| ELP4 | |
| Human recombit protein fragment corresponding to amino acids 133-407 of human ELP4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3A4 | |
| Unconjugated | |
| ELP4 | |
| AN, AN2, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26610 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction