missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELP4 Monoclonal Antibody (OTI3A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809150S
This item is not returnable.
View return policy
Description
ELP4 Monoclonal antibody specifically detects ELP4 in Human samples. It is validated for Western BlotSpecifications
| ELP4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96EB1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26610 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3A4 | |
| Unconjugated | |
| Elp4 | |
| A330107A17Rik; AN; C11orf19; dJ68P15A.1; elongation protein 4 homolog; elongation protein 4 homolog (S. cerevisiae); elongator acetyltransferase complex subunit 4; elongator complex protein 4; ELP4; hELP4; hypothetical protein LOC614139; PAX6 neighbor gene protein; PAX6NEB; PAXNEB; RGD1566052; similar to elongation protein 4 homolog | |
| Human recombinant protein fragment corresponding to amino acids 133-407 of human ELP4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction