Learn More
ELP4 Mouse anti-Human, Clone: OTI4H10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809167
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.Specifications
| ELP4 | |
| Monoclonal | |
| Unconjugated | |
| ELP4 | |
| AN, AN2, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26610 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4H10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96EB1 | |
| ELP4 | |
| Human recombit protein fragment corresponding to amino acids 133-407 of human ELP4 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.