missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELP4 Recombinant Rabbit Monoclonal Antibody (JE61-03)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544866
This item is not returnable.
View return policy
Description
ELP4 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
Specifications
| ELP4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q96EB1, Q9ER73 | |
| Elp4 | |
| Synthetic peptide within human Elp4 aa 313-364/424. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| JE61-03 | |
| Unconjugated | |
| Elp4 | |
| A330107A17Rik; AN; C11orf19; dJ68P15A.1; elongation protein 4 homolog; elongation protein 4 homolog (S. cerevisiae); elongator acetyltransferase complex subunit 4; elongator complex protein 4; ELP4; hELP4; hypothetical protein LOC614139; PAX6 neighbor gene protein; PAX6NEB; PAXNEB; RGD1566052; similar to elongation protein 4 homolog | |
| Rabbit | |
| Protein A | |
| RUO | |
| 26610, 77766 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction