missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELTD1/ETL Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS2113
This item is not returnable.
View return policy
Description
ELTD1/ETL Polyclonal antibody specifically detects ELTD1/ETL in Human, Mouse, Rat, Porcine, Bat, Chicken, Equine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ELTD1/ETL | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 4 - 11 μg/mL | |
| EGF, latrophilin and seven transmembrane domain containing 1, EGF, latrophilin and seven transmembrane domain-containing protein 1, ETL protein, ETLEGF-TM7-latrophilin-related protein, KPG_003 | |
| Synthetic 19 amino acid peptide from C-terminus of human ELTD1/ETL. | |
| 0.05 mL | |
| GPCR, Neuroscience | |
| 64123 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction