Learn More
ELTD1/ETL Antibody (CL4164) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59041AF647
Product Code. 30291108
  • € 737.00 / 0.10mL
Description
ELTD1/ETL Monoclonal antibody specifically detects ELTD1/ETL in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELTD1/ETL
Monoclonal
Alexa Fluor 647
EGF, latrophilin and seven transmembrane domain containing 1, EGF, latrophilin and seven transmembrane domain-containing protein 1, ETL protein, ETLEGF-TM7-latrophilin-related protein, KPG_003
This antibody was developed using a recombinant protein derived from Q9HBW9, with the exact immunogen sequence remaining proprietary.
0.1 mL
GPCR, Neuroscience
64123
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL4164
50mM Sodium Borate
Mouse
Protein A purified