missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Eltrombopag
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Eltrombopag is a thrombopoietin receptor (TpoR) agonist (EC50 = 0.27 μM in a reporter assay) and an iron chelator (Erickson‐Miller et al.) Interaction between Eltrombopag and TpoR triggers proliferation and differentiation of human bone marrow progenitor cells into megakaryocytes and increases platelet growth and production (Erickson‐Miller et al.)
- Stimulates proliferation and differentiation of primary human CD34+ bone marrow cells into CD41+ megakaryocytes (Erickson‐Miller et al.)
Specifications
Specifications
| CAS | 496775-61-2 |
| Synonym | SB497115 |
| Quantity | 1 mg |
| Molecular Formula | C25H22N4O4 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction