missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Eltrombopag
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Eltrombopag is a thrombopoietin receptor (TpoR) agonist (EC50 = 0.27 μM in a reporter assay) and an iron chelator (Erickson‐Miller et al.) Interaction between Eltrombopag and TpoR triggers proliferation and differentiation of human bone marrow progenitor cells into megakaryocytes and increases platelet growth and production (Erickson‐Miller et al.)
- Stimulates proliferation and differentiation of primary human CD34+ bone marrow cells into CD41+ megakaryocytes (Erickson‐Miller et al.)
Specifications
Specifications
| CAS | 496775-61-2 |
| Quantity | 5 mg |
| Molecular Formula | C25H22N4O4 |
| Purity | ≥98% |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion