Läs mer
Agilent BioTek ELx808 Qualification Package - IQ/OQ/PQ
Produktkod. 12501427 Handla allt< titel> Produkter
missing translation for 'orderingAttributeHoverText'
:
ELx808 Qualification Package - IQ/OQ/PQ
Denna artikel kan inte returneras. Se returpolicy
Note:
Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
Produktkod. 12501427
missing translation for 'mfr': Agilent BioTek 7340543
Denna artikel kan inte returneras. Se returpolicy
Note:
Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
For ELx808 microplate reader models
  • Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
  • Comes on USB flash drive with the following contents:
    • Detailed required material list
    • Installation qualification procedure and checklist
    • Operational qualification procedure
    • Performance qualification procedure
    • Microsoft Excel spreadsheets for calculations required by procedures
    • Maintenance procedures
    • 5 year logbook for documenting all procedures/results
      • All of the documentation is available in both .doc and .pdf file format
Note:
  • Absorbance test plate (7260522) may be required in order to run some procedures specified for absorbance and multi-mode readers, but is not included and must be ordered separately.
TRUSTED_SUSTAINABILITY
Specifikationer
For Use With (Equipment) ELx808 microplate reader models
Includes Detailed required material list, Installation qualification procedure and checklist, Operational qualification procedure, Performance qualification procedure, Microsoft Excel spreadsheets for calculations, Maintenance procedures, 5 year logbook
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.