missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMAP-II/AIMP1 Antibody (959507), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB19101UV
This item is not returnable.
View return policy
Description
EMAP-II/AIMP1 Monoclonal antibody specifically detects EMAP-II/AIMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry
Specifications
| EMAP-II/AIMP1 | |
| Monoclonal | |
| DyLight 350 | |
| aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43 | |
| E.coli-derived recombinant human EMAP-II, Ser147-Lys312, Accession # Q12904 | |
| 100 μg | |
| Cytokine Research | |
| 9255 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 959507 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction