Learn More
EMAP-II/AIMP1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98343AF532
Product Code. 18029869
  • € 415.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
EMAP-II/AIMP1 Polyclonal specifically detects EMAP-II/AIMP1 in Mouse samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
EMAP-II/AIMP1
Polyclonal
Western Blot, ELISA
aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Produced in rabbits immunized with purified, recombinant EMAP-II/AIMP1 (Ser145-Lys310).
0.1 mL
Biologically Active Proteins, Cytokine Research
9255
Store at 4°C in the dark.
IgG
Western Blot, ELISA
Alexa Fluor 532