missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMAP II Monoclonal Antibody (05)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA529014
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human AIMP1/EMAP II.
The protein encoded by this gene is a cytokine that is specifically induced by apoptosis, and it is involved in the control of angiogenesis, inflammation, and wound healing. The release of this cytokine renders the tumor-associated vasculature sensitive to tumor necrosis factor. The precursor protein is identical to the p43 subunit, which is associated with the multi-tRNA synthetase complex, and it modulates aminoacylation activity of tRNA synthetase in normal cells. This protein is also involved in the stimulation of inflammatory responses after proteolytic cleavage in tumor cells. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene has been identified on chromosome 20.
Specifications
| EMAP II | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q12904 | |
| Aimp1 | |
| Recombinant Human AIMP1/EMAP II Protein (Ala2-Lys312). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 05 | |
| Unconjugated | |
| Aimp1 | |
| 9830137A06Rik; aaRS-interacting multifunctional protein 1; AIMP1; AIMP1/p43; AIMP1-PA; Aminoacyl tRNA synthase complex-interacting multifunctional protein 1; aminoacyl tRNA synthetase complex interacting multifunctional protein 1; aminoacyl tRNA synthetase complex-interacting multifunctional protein 1; aminoacyl-tRNA synthetase-interacting multifunctional protein 1; ARS inducible multifunctional protein 1; ARS-interacting multifunctional protein 1; CG8235; CG8235-PA; Dmel\CG8235; Dmel_CG8235; EMAP2; EMAP-2; EMAPII; EMAP-II; endothelial monocyte activating polypeptide 2; endothelial monocyte-activating polypeptide 2; Endothelial monocyte-activating polypeptide II; endothelial-monocyte activating polypeptide II; HLD3; mca1; multisynthase complex auxiliary component p43; multisynthetase complex auxiliary component p43; p43; SCYE1; Small inducible cytokine subfamily E member 1; small inducible cytokine subfamily E, member 1; small inducible cytokine subfamily E, member 1 (endothelial monocyte-activating) | |
| Mouse | |
| Protein A | |
| RUO | |
| 9255 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction