missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Embigin Polyclonal Antibody
GREENER_CHOICE

Product Code. 16566251
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16566251 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16566251 Supplier Invitrogen™ Supplier No. PA587887

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DGSAPDSPFT SPPLREEIMA NNFSLESHNI SLTEHSSMPV EKNITLERPS NVNLTCQFTT SGDLNAVNVT WKKDGEQLEN NYLVSATGST LYTQYRFTII NSKQMGSYSC FFREEKEQRG TFNFKVPELH GKNKPLISYV GDSTVLTCKC QNCFPLNWTW YSSNGSVKVP VGVQMNKYVI NGTYANETKL KITQLLEEDG ESYWCRALFQ LGESEEHIEL VVLSYLVP; Positive Samples: Jurkat, MCF-7, THP-1; Cellular Location: Cell junction, Cell membrane, Single-pass type I membrane protein, synapse.

Embigin is a 327 amino acid regulatory protein belonging to the immunoglobulin superfamily class of CAMs, and has two Ig-like (immunoglobulin-like) V- type domains. In situ hybridization analysis shows that Embigin has specific carbohydrate epitopes and plays an important role in early postimplantation embryogenesis. A highly glycosylated transmembrane glycoprotein, Embigin acts as a regulator of cell/ECM interactions during development and homeostasis of normal adult tissues. Transfection studies suggest its role in promotion of integrin-mediated cell-substratum adhesion. Widely expressed in most of the tissues and embryonal carcinoma cells, prostate and mammary cancer cell lines, it may also help in the classification of human carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Embigin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EMB
Gene Accession No. Q6PCB8
Gene Alias AL022799; EMB; Embigin; embigin homolog; glycoprotein 70; Gp70; Teratocarcinoma glycoprotein Gp-70
Gene Symbols EMB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 33-260 of human EMB (NP_9408511).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133418
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.