missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMC1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18427AF647
This item is not returnable.
View return policy
Description
EMC1 Polyclonal antibody specifically detects EMC1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| EMC1 | |
| Polyclonal | |
| Western Blot | |
| hypothetical protein LOC23065, KIAA0090, RP1-43E13.1 | |
| Recognizes region between residues Between Between 875 and 925.. | |
| 0.1 mL | |
| Cell Biology | |
| 23065 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction