missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EMC4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA548708

Product Code. 15643005

  • € 520.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Predicted to react with bovine and mouse based on sequence homology.

May mediate anti-apoptotic activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EMC4
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q5J8M3
EMC4
KLH conjugated synthetic peptide between 34-62 amino acids from the N-terminal region of human TMM85.
400 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EMC4
2610318K02Rik; Cell proliferation-inducing gene 17 protein; EMC4; ER membrane protein complex subunit 4; HSPC184; PIG17; TMEM85; Transmembrane protein 85
Rabbit
Antigen affinity chromatography, Protein A
RUO
51234
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.