missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552688
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: DMRREMEQSM NMLNSNHELP DVSEFMTRLF SSKSSGKSSS GSSKTGKSGA GK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
EMC7 (ER Membrane Protein Complex Subunit 7) is a Protein Coding gene. Diseases associated with EMC7 include Cone-Rod Dystrophy, X-Linked, 1. Gene Ontology (GO) annotations related to this gene include carbohydrate binding. [GeneCards].
Specifikationer
| EMC7 | |
| Polyclonal | |
| Unconjugated | |
| EMC7 | |
| 2900064A13Rik; AI451465; c11orf3; C15orf24; chromosome 15 hypothetical ATG/GTP binding protein; Emc7; ER membrane protein complex subunit 7; HT022; ORF1-FL1; Orf3; UNQ905/PRO1926; UPF0480 protein C15orf24; UPF0480 protein C15orf24 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56851 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NPA0 | |
| EMC7 | |
| Recombinant protein corresponding to Human EMC7. Recombinant protein control fragment (Product #RP-90233). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering